Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcamr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170632.1 | NP_001164103.1 | 5 - 6 | 1548 | 68 | |
NM_144960.2 | NP_659209.2 | 5 - 6 | 1380 | 68 | ||
GenBank mRNA | AB048834.1 | - | 5 - 6 | 1392 | 68 | |
BC110549.1 | - | 5 - 6 | 1551 | 40047147 | 68 | |
BC110550.1 | - | 5 - 6 | 1383 | 40047153 | 68 | |
FJ011703.1 | - | 4 - 5 | 1461 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |