Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284411.1 | NP_001271340.1 | 3 - 4 | 325 | 103 | |
NM_001284412.1 | NP_001271341.1 | 3 - 4 | 301 | 103 | ||
NM_001284414.1 | NP_001271343.1 | 3 - 4 | 301 | 103 | ||
NM_026776.4 | NP_081052.2 | 3 - 4 | 301 | 103 | ||
GenBank mRNA | AK002500.1 | - | 3 - 4 | 263 | 103 | |
AK003715.2 | - | 3 - 4 | 273 | 103 | ||
AK003877.1 | - | 3 - 4 | 259 | 103 | ||
AK082933.1 | - | 3 - 4 | 280 | 103 | ||
AK145811.1 | - | 3 - 4 | 272 | 103 | ||
AK150822.1 | - | 3 - 4 | 283 | 103 | ||
AK162230.1 | - | 3 - 4 | 274 | 103 | ||
AK166008.1 | - | 3 - 4 | 280 | 103 | ||
AK190530.1 | - | 1 - 2 | 55 | 103 | ||
AK214794.1 | - | 1 - 2 | 55 | 103 | ||
BC029181.1 | - | 3 - 4 | 265 | 4240500 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |