Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181409.3 | NP_852074.2 | 13 - 14 | 1529 | 92 | |
GenBank mRNA | AK132375.1 | - | 13 - 14 | 1496 | 92 | |
BC051083.1 | - | 13 - 14 | 1462 | 6486043 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |