Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009957.1 | NP_034087.1 | 1 | 1044 | 88 | |
GenBank mRNA | AB008179.1 | - | 1 | 937 | 88 | |
AY013765.1 | - | 1 | 1044 | 88 | ||
BC080863.1 | - | 1 | 1166 | 30356469 | 88 | |
EU854476.1 | - | 1 | 918 | 88 |
Gene Symbol: Pcdha7-g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301259.1 | NP_001288188.1 | 1 | 1166 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |