Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007687.5 | NP_031713.1 | 2 - 3 | 451 | 90 | |
GenBank mRNA | AK013943.1 | - | 2 - 3 | 380 | 90 | |
AK020471.1 | - | 2 - 3 | 422 | 90 | ||
AK050651.1 | - | 382 | 90 | |||
AK088633.1 | - | 2 - 3 | 387 | 90 | ||
AK088793.1 | - | 2 - 3 | 386 | 90 | ||
AK167795.1 | - | 2 - 3 | 382 | 90 | ||
BC046225.1 | - | 2 - 3 | 346 | 5691650 | 90 | |
BC058726.1 | - | 2 - 3 | 379 | 5715707 | 90 | |
BC094357.1 | - | 2 - 3 | 361 | 6820355 | 90 | |
CX218255.1 | - | 2 - 3 | 487 | 90 | ||
D00472.1 | - | 2 - 3 | 400 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |