Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026991.2 | NP_081267.1 | 4 - 5 | 460 | 96 | |
GenBank mRNA | AK011406.1 | - | 5 - 6 | 379 | 96 | |
BC061227.1 | - | 4 - 5 | 460 | 30312881 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |