Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009011.4 | NP_033037.2 | 2 - 3 | 472 | 107 | |
GenBank mRNA | AK142280.1 | - | 2 - 3 | 464 | 107 | |
AK150089.1 | - | 2 - 3 | 464 | 107 | ||
AK157252.1 | - | 2 - 3 | 434 | 107 | ||
AK160880.1 | - | 2 - 3 | 464 | 107 | ||
AK160890.1 | - | 2 - 3 | 450 | 107 | ||
AK160973.1 | - | 2 - 3 | 450 | 107 | ||
AK167582.1 | - | 2 - 3 | 460 | 107 | ||
BC027747.1 | - | 2 - 3 | 461 | 4459657 | 107 | |
BC068193.1 | - | 2 - 3 | 431 | 6813997 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |