Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009011.4 | NP_033037.2 | 8 - 9 | 1266 | 100 | |
GenBank mRNA | AK142280.1 | - | 8 - 9 | 1258 | 100 | |
AK150089.1 | - | 8 - 9 | 1258 | 100 | ||
AK157252.1 | - | 8 - 9 | 1228 | 100 | ||
AK160880.1 | - | 8 - 9 | 1258 | 100 | ||
AK160890.1 | - | 8 - 9 | 1244 | 100 | ||
AK160973.1 | - | 8 - 9 | 1244 | 100 | ||
AK167582.1 | - | 8 - 9 | 1254 | 99 | ||
BC027747.1 | - | 8 - 9 | 1255 | 4459657 | 100 | |
BC068193.1 | - | 8 - 9 | 1222 | 6813997 | 100 | |
X92411.1 | - | 8 - 9 | 946 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |