Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Reck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016678.2 | NP_057887.2 | 13 - 14 | 1657 | 63 | |
GenBank mRNA | AB006960.1 | - | 13 - 14 | 1587 | 63 | |
AK042266.1 | - | 13 - 14 | 1659 | 63 | ||
AK143657.1 | - | 13 - 14 | 1636 | 63 | ||
AK157885.1 | - | 13 - 14 | 1645 | 63 | ||
BC053025.1 | - | 13 - 14 | 1596 | 63 | ||
BC132197.1 | - | 13 - 14 | 1685 | 40130474 | 63 | |
BC138065.1 | - | 13 - 14 | 1685 | 8861084 | 63 | |
BC145133.1 | - | 13 - 14 | 1686 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |