Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax6os1 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY589783.1 | - | LIT3401 Pax6OS |
2 - 3 | 142 | 112 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | reticulocalbin 1 | Rcn1 | Rcn | -224.4kb | Sense | Chr. 2 - 105385948 - 105399319 on NCBI Build GRCm38 |
Downstream | Pax6 upstream antisense RNA | Paupar | 33.0kb | Sense | Chr. 2 - 105657862 - 105661343 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |