Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Airn | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035282.1 | - | 1 | 1923 | 78 | ||
AU015441.1 | - | LIT1853 Air |
1 | 345 | 78 | ||
BG070272.2 | - | 1 | 345 | 78 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | insulin-like growth factor 2 receptor | Igf2r |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
-48.3kb | Anti-sense | Chr. 17 - 12682406 - 12769706 on NCBI Build GRCm38 |
Downstream | MAS1 oncogene | Mas1 |
Mas-1, Mgra |
20.5kb | Anti-sense | Chr. 17 - 12838841 - 12868145 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |