Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003247.2 | 2 - 3 | 145 | 62 | ||
GenBank mRNA | CK023612.1 | - | 143 | 62 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Rho GTPase activating protein 35 | Arhgap35 |
6430596G11Rik, AI841135, Grlf1, mKIAA1722, p190A, p190RhoGAP |
-34.0kb | Sense | Chr. 7 - 16494473 - 16614993 on NCBI Build GRCm38 |
Downstream | carcinoembryonic antigen-related cell adhesion molecule 15 | Ceacam15 |
AA407838, C430002N04Rik |
19.6kb | Sense | Chr. 7 - 16671331 - 16675705 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons |