Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042593.1 | NP_001036058.1 | 3 - 4 | 382 | 109 | |
NM_001145209.1 | NP_001138681.1 | 3 - 4 | 382 | 109 | ||
NM_019702.2 | NP_062676.2 | 3 - 4 | 382 | 109 | ||
GenBank mRNA | AK006626.1 | - | 3 - 4 | 355 | 109 | |
AK012856.1 | - | 3 - 4 | 332 | 109 | ||
AK146678.1 | - | 3 - 4 | 348 | 109 | ||
AK166140.1 | - | 3 - 4 | 345 | 109 | ||
AK168671.1 | - | 3 - 4 | 345 | 109 | ||
AK169281.1 | - | 3 - 4 | 345 | 109 | ||
AK173091.1 | - | 3 - 4 | 328 | 109 | ||
BC010251.1 | - | 3 - 4 | 307 | 3964560 | 109 | |
BC085235.1 | - | 3 - 4 | 324 | 5328213 | 109 | |
BU510509.1 | - | 3 - 4 | 334 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |