Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011614.3 | NP_035744.1 | 7 | 1058 | 101 | |
GenBank mRNA | AF030100.2 | - | 7 | 714 | 101 | |
AK020909.1 | - | 4 | 397 | 101 | ||
AK170297.1 | - | 7 | 958 | 101 | ||
AK171014.1 | - | 7 | 958 | 101 | ||
CB204861.1 | - | 2 | 336 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |