Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252629.1 | NP_001239558.1 | 8 - 9 | 967 | 106 | |
NM_011420.2 | NP_035550.1 | 8 - 9 | 903 | 106 | ||
GenBank mRNA | AI849087.1 | - | 1 - 2 | 106 | ||
AK144778.1 | - | 8 - 9 | 904 | 106 | ||
AK152495.1 | - | 8 - 9 | 877 | 106 | ||
AK167832.1 | - | 8 - 9 | 901 | 106 | ||
AY821787.1 | - | 5 - 6 | 272 | 106 | ||
BC037436.1 | - | 3 - 4 | 352 | 106 | ||
BC045158.1 | - | 8 - 9 | 867 | 6486685 | 106 | |
EU234021.1 | - | 8 - 9 | 967 | 106 | ||
U63294.1 | - | 8 - 9 | 881 | 106 | ||
U77714.1 | - | 8 - 9 | 864 | 106 | ||
Y12835.1 | - | 8 - 9 | 889 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |