Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncald | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033226.1 | - | 2 - 3 | 406 | 70 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930447A16 gene | 4930447A16Rik | -178.1kb | Anti-sense | Chr. 15 - 37423867 - 37440644 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 35019 |
Gm35019 | 231.9kb | Sense | Chr. 15 - 37893003 - 37907777 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |