Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033279.1 | - | 1 | 1851 | 82 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930452A19 gene | 4930452A19Rik | 4933424P20Rik | -426.7kb | Sense | Chr. 17 - 9750752 - 9775866 on NCBI Build GRCm38 |
Downstream | colon adenocarcinoma hypermethylated RNA | Cahm | B930003M22Rik | 115.0kb | Anti-sense | Chr. 17 - 10319919 - 10321141 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |