Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020100.1 | - | 1 | 347 | 80 | |
AK082054.1 | - | 1 | 352 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NSA2 ribosome biogenesis homolog (S. cerevisiae) | Nsa2 |
5730427N09Rik, LNR42, Nsa2p, Tinp1, Yr29 |
-72.0 | Anti-sense | Chr. 13 - 97129427 - 97137926 on NCBI Build GRCm38 |
Downstream | hexosaminidase B | Hexb | 37.9kb | Anti-sense | Chr. 13 - 97176331 - 97198357 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |