Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm12758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK087850.1 | - | 117 | 82 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4931406P16 gene | 4931406P16Rik |
AA553327, mKIAA0355 |
Anti-sense | Chr. 7 - 34236707 - 34313575 on NCBI Build GRCm38 | |
Downstream |
LSM14 homolog A (SCD6, S. cerevisiae) |
Lsm14a |
2700023B17Rik, AA407828, AU017544, Tral |
107.9kb | Anti-sense | Chr. 7 - 34344637 - 34389637 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |