Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK080243.1 | - | 1 | 366 | 122 | |
AK088236.1 | - | 1 | 346 | 122 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ubiquitin A-52 residue ribosomal protein fusion product 1 pseudogene | Gm11517 | OTTMUSG00000001634 | -9.1kb | Anti-sense | Chr. 11 - 96795332 - 96798156 on NCBI Build GRCm38 |
Downstream |
nuclear factor, erythroid derived 2,-like 1 |
Nfe2l1 |
AA408798, AW212678, LCR-F1, Lcrf1, NRF1, TCF-11, TCF11 |
9.3kb | Anti-sense | Chr. 11 - 96817413 - 96829968 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |