Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138215.1 | - | 1 - 2 | 130 | 75 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
DnaJ (Hsp40) homolog, subfamily C, member 24 |
Dnajc24 |
1700030A21Rik, 2610027M02Rik, AV066965, AW240712, Dph4, MmDjC7, Zcsl3 |
-222.6kb | Anti-sense | Chr. 2 - 105966708 - 106003868 on NCBI Build GRCm38 |
Downstream | predicted gene 14015 | Gm14015 |
9330126M15, OTTMUSG00000015366 |
234.3kb | Anti-sense | Chr. 2 - 106463823 - 106523103 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |