Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK157936.1 | - | 1 - 2 | 1132 | 77 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
tandem C2 domains, nuclear |
Tc2n |
4933406D09Rik, Mtac2d1, Tac2-N |
-97.4kb | Sense | Chr. 12 - 101645446 - 101719000 on NCBI Build GRCm38 |
Downstream | thyroid hormone receptor interactor 11 | Trip11 |
2610511G22Rik, 3110031G15Rik, 6030460N08Rik, AI450776, GMAP-210, TRIP230 |
15.0kb | Sense | Chr. 12 - 101834043 - 101913649 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |