Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vti1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK003692.1 | - | 1 | 454 | 101 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
zinc finger, DHHC domain containing 6 |
Zdhhc6 |
2400007G07Rik, 5930409M18Rik, AI462345, C77369 |
-131.8kb | Anti-sense | Chr. 19 - 55298293 - 55316032 on NCBI Build GRCm38 |
Downstream | predicted gene 4466 | Gm4466 | 33.7kb | Anti-sense | Chr. 19 - 55482087 - 55487437 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |