Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632418H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019501.1 | - | 1 | 2046 | 108 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SNF related kinase | Snrk |
2010012F07Rik, AI448042, AW547029, E030034B15, R74830, mKIAA0096 |
Sense | Chr. 9 - 122117266 - 122169707 on NCBI Build GRCm38 | |
Downstream | anoctamin 10 | Ano10 |
AI604832, Tmem16k |
45.9kb | Anti-sense | Chr. 9 - 122175874 - 122294443 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |