Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | T cell receptor alpha chain | Tcra | Tcralpha | Sense | Chr. 14 - 52427967 - 54224198 on NCBI Build GRCm38 | |
Downstream | defender against cell death 1 | Dad1 | AI323713 | 58.0kb | Anti-sense | Chr. 14 - 54235485 - 54253929 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |