Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: l7Rn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111916.1 | 1 | 1689 | 97 | ||
GenBank mRNA | AK133534.1 | - | 1 | 1648 | 97 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 81 | Ccdc81 | 4921513D09Rik | -35.0kb | Sense | Chr. 7 - 89866148 - 89903629 on NCBI Build GRCm38 |
Downstream | embryonic ectoderm development | Eed |
ENSMUSG00000039373, l(7)5Rn, l7Rn5, lusk |
13.5kb | Sense | Chr. 7 - 89954654 - 89981419 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |