Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK053535.1 | - | 1 | 3648 | 82 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 397 | Zfp397 |
2810411K16Rik, 6720480F11Rik, AI461664 |
-18.6kb | Anti-sense | Chr. 18 - 23954662 - 23964671 on NCBI Build GRCm38 |
Downstream | zinc finger protein 35 | Zfp35 |
Zfp-35, Znf271, mszf13, mszf20-1, mszf28, mszf68 |
2.4kb | Anti-sense | Chr. 18 - 23989634 - 24005371 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |