Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | procollagen C-endopeptidase enhancer protein | Pcolce |
Astt-2, Astt2, P14, PCPE-1 |
-955.0 | Anti-sense | Chr. 5 - 137605105 - 137611899 on NCBI Build GRCm38 |
Downstream | predicted gene 20605 | Gm20605 | Lrch4-sap25 | 15.0kb | Sense | Chr. 5 - 137629123 - 137642896 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |