Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 3473f | Mir3473f |
Gm27846, mmu-mir-3473f |
-132.6kb | Sense | Chr. 1 - 106546496 - 106546631 on NCBI Build GRCm38 |
Downstream | 3-ketodihydrosphingosine reductase | Kdsr |
6330410P18Rik, 9430079B08Rik, Fvt1 |
40.5kb | Sense | Chr. 1 - 106720410 - 106759830 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |