Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
cytochrome P450, family 4, subfamily a, polypeptide 14 |
Cyp4a14 | AI314743 | Anti-sense | Chr. 4 - 115486200 - 115496159 on NCBI Build GRCm38 | |
Downstream |
cytochrome P450, family 4, subfamily a, polypeptide 10 |
Cyp4a10 |
AI647584, Cyp4a, D4Rp1, RP1 |
23.0kb | Sense | Chr. 4 - 115518287 - 115533649 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |