Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
platelet-activating factor acetylhydrolase, isoform 1b, subunit 1 |
Pafah1b1 |
LIS-1, Lis1, MMS10-U, Mdsh, Ms10u, Pafaha |
Sense | Chr. 11 - 74673949 - 74724384 on NCBI Build GRCm38 | |
Downstream | methyltransferase like 16 | Mettl16 |
2610100D03Rik, 2810013M15Rik, A830095F14Rik, AI846133, Mett10d |
78.6kb | Anti-sense | Chr. 11 - 74770836 - 74818694 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |