Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK161004.1 | - | 3 | 2551 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | poly(rC) binding protein 2 | Pcbp2 |
AW412548, Hnrpx, alphaCP-2 |
-6.1kb | Anti-sense | Chr. 15 - 102470222 - 102500059 on NCBI Build GRCm38 |
Downstream | TAR (HIV) RNA binding protein 2 | Tarbp2 |
Prbp, TRBP |
8.8kb | Anti-sense | Chr. 15 - 102518192 - 102523676 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |