Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zar1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159693.1 | NP_001153165.1 | 1 - 2 | 570 | 91 | |
GenBank mRNA | FJ858201.1 | - | 1 - 2 | 570 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |