Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800003465.mm.1 |
NM_001160412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003465.mm.1 |
NM_001160412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003465.mm.1 |
TC0800000501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800003463.mm.1 |
NM_001160412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003463.mm.1 |
NM_001160412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003463.mm.1 |
TC0800000501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800003466.mm.1 |
NM_001160412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003466.mm.1 |
NM_001160412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003466.mm.1 |
TC0800000501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800003464.mm.1 |
NM_001160412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003464.mm.1 |
NM_001160412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003464.mm.1 |
TC0800000501.mm.1 |
MTA TC |