Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160424.1 | NP_001153896.1 | 2 - 3 | 439 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |