Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161742.1 | NP_001155214.1 | 2 - 3 | 178 | 81 | |
NM_001161744.1 | NP_001155216.1 | 1 - 2 | 117 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |