Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6g6f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163192.1 | NP_001156664.1 | 4 - 5 | 831 | 94 | |
GenBank mRNA | BC157908.1 | - | 4 - 5 | 831 | 9088110 | 94 |
BC172069.1 | - | 4 - 5 | 831 | 9054355 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |