Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163317.1 | NP_001156789.1 | 2 - 3 | 497 | 58 | |
GenBank mRNA | BC147370.1 | - | 2 - 3 | 496 | 9056434 | 58 |
BC147371.1 | - | 2 - 3 | 496 | 9056432 | 58 | |
BC147726.1 | - | 2 - 3 | 497 | 9056675 | 58 | |
BC147731.1 | - | 2 - 3 | 497 | 9056680 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |