Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700002923.mm.1 |
NM_001163416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002923.mm.1 |
NM_001163416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002923.mm.1 |
NM_026747.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002923.mm.1 |
NM_026747.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002923.mm.1 |
TC1700000361.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002926.mm.1 |
NM_001163416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002926.mm.1 |
NM_001163416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002926.mm.1 |
NM_026747.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002926.mm.1 |
NM_026747.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002926.mm.1 |
TC1700000361.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002925.mm.1 |
NM_001163416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002925.mm.1 |
NM_001163416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002925.mm.1 |
NM_026747.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002925.mm.1 |
NM_026747.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002925.mm.1 |
TC1700000361.mm.1 |
MTA TC |