Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164201.1 | NP_001157673.1 | 8 - 9 | 980 | 93 | |
GenBank mRNA | DQ358087.1 | - | 7 - 8 | 738 | 93 | |
DQ646881.1 | - | 8 - 9 | 859 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |