Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10421397 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421398 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421411 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421403 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421416 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421415 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421404 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421402 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421409 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421395 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421400 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421399 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421414 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421410 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421417 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421413 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421407 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421405 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421406 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421408 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421412 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421396 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10421401 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1457246_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10421394 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10416332 |
- |
- |
Mouse Genome 430 2.0 Array |
1457246_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1457246_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
239170_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
239170_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1400018320.mm.1 |
NM_194345.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018320.mm.1 |
NM_194345.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018320.mm.1 |
TC1400002345.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1457246_PM_at |
- |
- |
Mouse Expression Set 430 |
1457246_at |
- |
- |
Murine Genome U74v2 Set |
129075_PM_at |
- |
- |
Murine Genome U74v2 Set |
129075_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1457246_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1457246_at |
- |
- |