Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800012276.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012276.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012276.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012273.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012273.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012273.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012283.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012283.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012283.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012284.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012284.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012284.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012275.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012275.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012275.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012281.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012281.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012281.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012277.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012277.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012277.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012282.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012282.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012282.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012274.mm.1 |
NM_018821.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012274.mm.1 |
NM_018821.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012274.mm.1 |
TC1800001728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012278.mm.1 |
TC1800001728.mm.1 |
MTA TC |