Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400026278.mm.1 |
TC0400003296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026278.mm.1 |
NM_001167876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026278.mm.1 |
NM_001167876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026278.mm.1 |
NM_001167878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026278.mm.1 |
NM_001167878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026279.mm.1 |
TC0400003296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026279.mm.1 |
NM_001167876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026279.mm.1 |
NM_001167876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026279.mm.1 |
NM_001167878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026279.mm.1 |
NM_001167878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026276.mm.1 |
TC0400003296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026276.mm.1 |
NM_001167876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026276.mm.1 |
NM_001167876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026276.mm.1 |
NM_001167878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026276.mm.1 |
NM_001167878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026277.mm.1 |
TC0400003296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026277.mm.1 |
NM_001167876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026277.mm.1 |
NM_001167876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026277.mm.1 |
NM_001167878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026277.mm.1 |
NM_001167878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026281.mm.1 |
TC0400003296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026281.mm.1 |
NM_001167876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026281.mm.1 |
NM_001167876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026281.mm.1 |
NM_001167878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026281.mm.1 |
NM_001167878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026280.mm.1 |
TC0400003296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026280.mm.1 |
NM_001167876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026280.mm.1 |
NM_001167876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026280.mm.1 |
NM_001167878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026280.mm.1 |
NM_001167878.1 |
Refseq |