Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Skint5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167876.1 | NP_001161348.1 | 5 - 6 | 858 | 131 | |
NM_001167878.1 | NP_001161350.1 | 5 - 6 | 858 | 131 | ||
GenBank mRNA | EU099301.1 | - | 5 - 6 | 858 | 131 | |
EU099302.1 | - | 5 - 6 | 858 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |