Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004159.2 | NP_001004159.1 | 3 - 4 | 244 | 106 | |
GenBank mRNA | AK156711.1 | - | 3 - 4 | 376 | 106 | |
AY365133.1 | - | 3 - 4 | 267 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |