Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10442038 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442042 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442043 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442041 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442038 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442043 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442044 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442045 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442042 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442044 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442040 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442040 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442041 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10442045 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10442037 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10442037 |
- |
- |
Mouse Genome 430 2.0 Array |
1449972_PM_s_at |
- |
- |
Mouse Genome 430 2.0 Array |
1449972_PM_s_at |
- |
- |
Mouse Genome 430 2.0 Array |
1449972_s_at |
- |
- |
Mouse Genome 430 2.0 Array |
1449972_s_at |
- |
- |
Affymetrix Murine Genome 6500A Array |
Msa14830_r_at |
- |
- |
Affymetrix Murine Genome 6500A Array |
Msa14830_i_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1700001605.mm.1 |
NM_011765.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001605.mm.1 |
NM_011765.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001605.mm.1 |
TC1700000198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001603.mm.1 |
NM_011765.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001603.mm.1 |
NM_011765.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001603.mm.1 |
TC1700000198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001588.mm.1 |
NM_001005358.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001588.mm.1 |
NM_001005358.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001588.mm.1 |
NM_001163919.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001588.mm.1 |
NM_001163919.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001588.mm.1 |
TC1700000194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001592.mm.1 |
NM_001005358.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001592.mm.1 |
NM_001005358.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001592.mm.1 |
NM_001163919.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001592.mm.1 |
NM_001163919.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001592.mm.1 |
TC1700000194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001607.mm.1 |
NM_011765.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001607.mm.1 |
NM_011765.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001607.mm.1 |
TC1700000198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001590.mm.1 |
NM_001005358.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001590.mm.1 |
NM_001005358.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001590.mm.1 |
TC1700000194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001606.mm.1 |
NM_011765.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001606.mm.1 |
NM_011765.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001606.mm.1 |
TC1700000198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001604.mm.1 |
NM_011765.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001604.mm.1 |
NM_011765.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001604.mm.1 |
TC1700000198.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1449972_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1449972_s_at |
- |
- |
Mouse Expression Set 430 |
1449972_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1449972_s_at |
- |
- |
Mouse Expression Set 430 |
1442940_PM_at |
- |
- |
Mouse Expression Set 430 |
1442940_at |
- |
- |
Murine Genome U74v2 Set |
98762_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
98762_f_at |
- |
- |
Murine Genome U74v2 Set |
98761_i_at |
- |
- |
Murine Genome U74v2 Set |
98761_i_at |
- |
- |
Murine Genome U74v2 Set |
98761_PM_i_at |
- |
- |
Murine Genome U74v2 Set |
98761_PM_i_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449972_PM_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449972_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449972_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449972_PM_s_at |
- |
- |