Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100034427.mm.1 |
NM_016880.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034427.mm.1 |
NM_016880.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034427.mm.1 |
TC1100003753.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034432.mm.1 |
NM_016880.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034432.mm.1 |
NM_016880.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034432.mm.1 |
TC1100003753.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034433.mm.1 |
NM_016880.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034433.mm.1 |
NM_016880.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034433.mm.1 |
TC1100003753.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034430.mm.1 |
NM_016880.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034430.mm.1 |
NM_016880.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034430.mm.1 |
TC1100003753.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034431.mm.1 |
NM_016880.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034431.mm.1 |
NM_016880.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034431.mm.1 |
TC1100003753.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034429.mm.1 |
NM_016880.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034429.mm.1 |
NM_016880.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034429.mm.1 |
TC1100003753.mm.1 |
MTA TC |