Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019946.4 | NP_064330.2 | 1 - 2 | 68 | 111 | |
GenBank mRNA | AK005122.1 | - | 1 - 2 | 69 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |