Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019969.3 | NP_064353.2 | 4 - 5 | 481 | 87 | |
GenBank mRNA | AY574219.1 | - | 4 - 5 | 458 | 87 | |
BC139283.1 | - | 4 - 5 | 431 | 8862305 | 87 | |
BC139285.1 | - | 4 - 5 | 431 | 8862307 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |