Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025083.2 | NP_001020254.1 | 1 - 2 | 245 | 110 | |
GenBank mRNA | BC132522.1 | - | 1 - 2 | 240 | 40130799 | 110 |
BC132548.1 | - | 1 - 2 | 240 | 40130825 | 110 | |
DQ058649.1 | - | 1 - 2 | 245 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |